missing translation for 'onlineSavingsMsg'
Learn More

Kinase Quench Buffer

Catalog No. P2825
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2825 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2825 Supplier Thermo Fisher Scientific Supplier No. P2825

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kinase Quench Buffer

Kinase Quench Buffer (500 mM EDTA) is an essential component for kinase assays using LanthaScreen™ technology. The Kinase Quench Buffer is used to stop the kinase reaction.

Specifications

Chemical Name or Material Kinase Quench Buffer
Physical Form Liquid
Quantity 1 mL
Includes Kinase Quench Buffer (500 mM EDTA in water) may be is shipped frozen or at room temperature. Store at room temperature (20-30°C).

For Research Use Only. Not for use in diagnostic procedures.