missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kinase Buffer S (5X)
Description
Kinase buffer that has been specially optimized for use with LRRK2 and LRRK2 mutants.
Specifications
Specifications
| Chemical Name or Material | Kinase Buffer S |
| Concentration | 5X |
| For Use With (Equipment) | LRRK2 and LRRK2 mutants |
| Physical Form | Liquid |
| Quantity | 4 mL |
| Includes | 5X Kinase Buffer S may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C). |
For Research Use Only. Not for use in diagnostic procedures.