missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Kimtech™ Prep Wipers

Designed for use in WetTask™ Refillable Wet Wiping System

$88.32 - $151.09

Specifications

Product Type Critical Task Wipe
Color White
Disposable Single-use
Format Roll
For Use With (Equipment) WetTask™ refillable wet wiping system
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Material Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Material Packaging Quantity Price Quantity & Availability  
29634009
View Documents Promotion Details
Kimberly-Clark Professional™
06211
12.5 x 12 in. Spunlace 90 Sheets/Roll
Case of 540 Each for $151.09
Only null left
 
19168391
View Documents Promotion Details
Kimberly-Clark Professional™
06471
12.5 x 12 in. Meltblown 140 Sheets/Roll
Case of 6 Each for $104.06
Only null left
 
296361449
View Documents Promotion Details
Kimberly-Clark Professional™
06001
12.5 x 12 in. Non-woven Polyproylene Cellulose 60 Sheets/Roll
Case of 6 Each for $88.32
Only null left
 
191312700
View Documents Promotion Details
Kimberly-Clark Professional™
77320
12.5 x 12 in. Meltblown 35 Sheets/Roll
Case of 12 Each for $89.79
Only null left
 
191302378
View Documents Promotion Details
Kimberly-Clark Professional™
06411
12.5 x 12 in. Meltblown 90 Sheets/Roll
Case of 540 Each for $111.66
Only null left
 
Description

Description

  • System is an ideal alternative to open buckets and rags; reduces exposure to chemical vapors and splashes
  • Enclosed system helps avoid contamination of wipers and cleaning solution
  • Maintains concentration of actives released at near target levels over a prolonged period of use

Specifications

Specifications

Critical Task Wipe
Single-use
WetTask™ refillable wet wiping system
White
Roll
Videos
SDS
Documents

Documents