Learn More
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns
Resist attack by liquids or vapors
$901.43 - $1,265.86
Specifications
Material Borosilicate Glass
View More Specs
Products 4
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
103101
View Documents Promotion Details
DWK Life Sciences
135003
3 mm
Case for $1,265.86
Only null left
103102
View Documents Promotion Details
DWK Life Sciences
135004
4 mm
Case for $1,192.39
Only null left
103103
View Documents Promotion Details
DWK Life Sciences
135005
5 mm
Case for $901.43
Only null left
103104
View Documents Promotion Details
DWK Life Sciences
135006
6 mm
Case for $917.71
Only null left
Description
Description
May be used as mixing beads, boiling stones, or packing
Order Info
Packed in 1 lb. boxes
Specifications
Specifications
Borosilicate Glass