Learn More
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns
Resist attack by liquids or vapors
Supplier:  DWK Life Sciences 135005
 View more versions of this product
Catalog No. 103103
Only null left
Description
Description
May be used as mixing beads, boiling stones, or packing
Order Info
Packed in 1 lb. boxes
Specifications
Chemical Identifiers
Borosilicate Glass
Specifications
Borosilicate Glass
5 mm
Product Suggestions
Product Suggestions