missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Serum KG-35 Borosilicate Glass Aluminum Seal without Closure

Catalog No. p-307150
Change view
Click to view available options
Capacity (Metric):
5 mL
50 mL
100 mL
125 mL
200 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
10310240J 5 mL
10310240K 50 mL
10310240B 100 mL
10310240C 125 mL
10310240F 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10310240J Supplier DWK Life Sciences Supplier No. 61000G5
Only null left

Neutraglas™

  • No closure
  • GPI Neck Finish: 20A

Specifications

Capacity 0.16 oz., 5mL
Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Diameter (English) 0.9 in.
Material Borosilicate Glass
Length (Metric) 47mm
Product Type Serum Bottle
Length (English) 1.85 in.
Closure Material Aluminum
Closure Size 20A
Diameter (Metric) 23 mm
Show More Show Less