missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Sample Vials Screw Thread KG-33 Borosilicate Glass
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
$154.64 - $1,883.75
Specifications
| Classification | A |
|---|---|
| Material (Liner) | Rubber |
| Color | Clear |
| Material | Borosilicate Glass |
| Thread Style | Screw |
| missing translation for 'productCode' | missing translation for 'brand' | Certifications/Compliance | Volume (English) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Certifications/Compliance | Volume (English) | Price | Quantity & Availability | ||||
|
0334043
|
DWK Life Sciences
60940D1 |
ASTM E438, Type I, Class A | 1 dr. |
Pack of 144 Each for $154.64
Case of 16 Pack for $2,474.24
|
|
||||
|
0334045
|
DWK Life Sciences
60940D3 |
ASTM Type I, Class A | 3.2 dr. |
Case of 1152 Each for $1,883.75
|
|
||||
|
0334044
|
DWK Life Sciences
60940D2 |
ASTM E438, Type I, Class A | 2.16 dr. |
Pack of 144 Each for $241.25
Case of 12 Pack for $2,895.00
|
|
||||
|
0334046
|
DWK Life Sciences
60940D4 |
ASTM Type I, Class A | 4.32 dr. |
Pack of 144 Each for $281.46
Case of 8 Pack for $2,251.68
|
|
||||
|
0334047
|
DWK Life Sciences
60940D6 |
ASTM Type I, Class A | 6.5 dr. |
Case of 864 Each for $1,767.53
|
|
||||
Description
- Glass sample vial includes unattached rubber-lined black phenolic closure
- Vial top has a screw thread finish
- Kimax KG-33 borosilicate glass - meets the requirements for ASTM Type 1, Class A and USP Type 1 Glass
- High chemical resistance
Specifications
| A | |
| Clear | |
| Screw | |
| Screw Cap (Closed-top) |
| Rubber | |
| Borosilicate Glass | |
| Phenolic |