Learn More
DWK Life Sciences Kimble™ Sample Vials Screw Thread KG-33 Borosilicate Glass
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
Supplier:  DWK Life Sciences 60940D1
 View more versions of this product
Catalog No. 0334043
Only null left
Description
Description
  • Glass sample vial includes unattached rubber-lined black phenolic closure
  • Vial top has a screw thread finish
  • Kimax KG-33 borosilicate glass - meets the requirements for ASTM Type 1, Class A and USP Type 1 Glass
  • High chemical resistance
Specifications
Chemical Identifiers
A
Clear
Screw
Screw Cap (Closed-top)
Rubber
Borosilicate Glass
Phenolic
Specifications
A
Clear
Borosilicate Glass
Screw
Phenolic
ASTM E438, Type I, Class A
Rubber
15 mm
4 mL
1 dr.
Screw Cap (Closed-top)
45 mm
Product Suggestions
Product Suggestions