missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Safety System Three Kit
Reduces the safety hazards of aluminum-foil wrapped solvent containers
$827.29 - $1,262.04
Specifications
| No. per Case | 1 Cs. |
|---|---|
| Thread Size (Metric) | GL 45 |
| For Use With (Equipment) | HPLC pump |
Includes:
- One three-hole cap, GL-45 with TFE/polypropylene O-ring and polypropylene screw collar
- One PBT solvent bottle adapter (GL-45) that converts the three-hole cap to fit standard 4L bottles
- One 2μm inlet/sparge filter with connections for 1/8 in. O.D. tubing
- Two flangeless ETFE nuts, 1/4-28 x 1/8 in., two flangeless ETFE ferrules, 1/8 in.
- One ETFE vent port plug, 1/4-28
- FEP PTFE tubing, 1/16 in. I.D. x 1/8 in. O.D. x 10 ft.
Description
- Inlet/sparger filter's all-PEEK™ construction excellent for virtually all mobile phases
Specifications
| 1 Cs. | |
| GL 45 | |
| HPLC pump |