Learn More
DWK Life Sciences Kimble™ Safety System Three Kit
Catalog No. 11301236
Change view
Click to view available options
Includes:
GL45 Bottle Adapter
Without GL45 Bottle Adapter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
11301236 Without GL45 Bottle Adapter
0642155 GL45 Bottle Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11301236 Supplier DWK Life Sciences Supplier No. 9539300001
Includes
  • One three-hole cap, GL-45 with TFE/polypropylene O-ring and polypropylene screw collar
  • One PBT solvent bottle adapter (GL-45) that converts the three-hole cap to fit standard 4L bottles
  • One 2μm inlet/sparge filter with connections for 1/8 in. O.D. tubing
  • Two flangeless ETFE nuts, 1/4-28 x 1/8 in., two flangeless ETFE ferrules, 1/8 in.
  • One ETFE vent port plug, 1/4-28
  • FEP PTFE tubing, 1/16 in. I.D. x 1/8 in. O.D. x 10 ft.
Reduces the safety hazards of aluminum-foil wrapped solvent containers
  • Inlet/sparger filter's all-PEEK™ construction excellent for virtually all mobile phases
Specifications
Includes Without GL45 Bottle Adapter
No. per Case 1 Cs.
Thread Size (Metric) GL 45
For Use With (Equipment) HPLC pump