missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Two Neck Round Bottom Heavy Wall Distilling Flasks
Outer joints. Side neck is angled.
Supplier: DWK Life Sciences 2949900050
Description
- Heavy wall reduces breakage.
Specifications
| 1.8 in. | |
| Borosilicate Glass | |
| Angled | |
| 14/20 Center, 14/20 side | |
| 48 mm |
| 50 mL | |
| Standard taper outer joint | |
| 2 | |
| Round |