missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Round Bottom Four Neck Heavy Wall Distilling Flasks
With vertical outer joints
Supplier: DWK Life Sciences 6070001124
Description
- Heavy wall reduces breakage.
Specifications
| 6.33 in. | |
| Borosilicate Glass | |
| Vertical | |
| 29/42 Center, 24/40 side | |
| 160 mm |
| 2000 mL | |
| Standard taper outer joint | |
| 4 | |
| Round |