missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Round Bottom Four Neck Heavy Wall Distilling Flasks
With vertical outer joints
Supplier: DWK Life Sciences 6070000924
Description
- Heavy wall reduces breakage.
Specifications
| 5.11 in. | |
| Borosilicate Glass | |
| Vertical | |
| 45/50 Center, 24/40 side |
| 1000 mL | |
| Standard taper outer joint | |
| 4 | |
| Round |