Learn More
DWK Life Sciences KIMBLE™ KONTES™ Replacement Plugs for Hi-Vac Right Angle Valve with Tip O-Ring
Catalog No. K8265010008
Change view
Click to view available options
Plug Stopcock No:
4
8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Stopcock No
K8265010008 8
K8265010004 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K8265010008 Supplier DWK Life Sciences Supplier No. 8265010008
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KIMBLE™ KONTES™ Replacement Plugs for Hi-Vac Right Angle Valve with Tip O-Ring are PTFE valve plugs designed for use in high-vacuum applications. Compatible with HI-VAC Right-Angle valves, these replacement plugs help maintain reliable vacuum sealing performance.
  • PTFE construction for chemical resistance and reliable sealing in high-vacuum applications
  • Designed as replacement plugs for Kimble Kontes HI-VAC Right-Angle valves with Tip O-Ring
  • Compatible with HI-VAC Right-Angle valves rated for vacuum applications up to 5 x 10 (8315 and 8311) Torr
  • Available in multiple sizes to match corresponding valve stem bore ranges
  • Sold individually (Case Qty: 1)
Specifications
Actuation Manual
Volume (Metric) Sample Size 8
For Use With (Equipment) HI-Vac Right Angle Valve
Material PTFE
Plug Stopcock No 8
Type Metering High Vacuum Valve Plug
Stator Material PTFE/CTFE