Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Septum Sample Tube Caps
For 5mm, 7mm and 10mm NMR Tubes
Supplier:  DWK Life Sciences 8970980007
 View more versions of this product
Catalog No. K8970980007
Only null left
Description
Description
For 5mm, 7mm and 10mm NMR Tubes
Soft, natural rubber allows easy penetration with a syringe needle
  • Dual seal, inside tube and outer sleeve
  • Ideal for thin wall tubes
  • Provides penetration point for syringe or cannulation additions
  • Useful as a closure or for sample outgassing
Specifications
Specifications
Septa
Red
7 mm Thin-walled NMR Tube
Rubber
Provides penetration point for syringe or cannulation additions and useful as a closure or for sample outgassing
Product Suggestions
Product Suggestions