Learn More
DWK Life Sciences KIMBLE™ KONTES™ MIDI-VAP 4000 Distillation Glassware Screw Cap
Catalog No. K4104790014
Change view
Click to view available options
Size:
GL 14
Catalog No. Size
K4104790014 GL 14
1 options
Catalog No. K4104790014 Supplier DWK Life Sciences Supplier No. 4104790014
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Genuine replacement and optional components for the MIDI-VAP 4000 Distillation Apparatus, supporting ammonia, phenol, and cyanide distillation workflows.
Component parts for both ammonia and phenol distillation system
  • Genuine replacement and optional parts for the Kimble™ Kontes™ MIDI-VAP 4000 Distillation Apparatus
  • Supports ammonia, phenol, and cyanide distillation applications
  • Includes glassware sets, tubing kits, reaction tubes, distilling heads, caps, septa, sealing rings, quick disconnects, and manifold heater
  • Borosilicate glass components designed for reduced sample volume semi-automatic colorimetry methods
  • Compatible with drinking, ground, surface, and saline water analysis as well as domestic and industrial waste testing
Specifications
For Use With (Application) Distillation Systems
Type Screw Cap
Material PTFE/Silicone Liner
For Use With (Equipment) MIDI-VAP 4000 Ammonia/Phenol System
Size GL 14