Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders
Combined conical and cylindrical surfaces improve grinding efficiency
$477.07 - $910.79
Specifications
Clearance (English) 0.004 to 0.006 in.
Clearance (Metric) 0.1 to 0.15mm
For Use With (Application) Ideal for connective tissue such as muscle, heart and lung
View More Specs
Products 11
Catalog Number Mfr. No. Capacity Includes Size Price Quantity  
Catalog Number Mfr. No. Capacity Includes Size Price Quantity  
K885480-0021
View Documents
DWK Life Sciences
8854800021
0.1 oz., 3mL PTFE pestle and Glass tube 21
Each for $477.07
Only null left
K885450-0020
View Documents
DWK Life Sciences
8854500020
0.03 oz., 1mL Ground Glass pestle and tube 20
Case for $661.89
Only null left
K885450-0024
View Documents
DWK Life Sciences
8854500024
1.18 oz., 30mL Ground Glass pestle and tube 24
Each for $717.54
Only null left
K885480-0023
View Documents
DWK Life Sciences
8854800023
0.59 oz., 15mL PTFE pestle and Glass tube 23
Each for $609.68
Only null left
K885450-0022
View Documents
DWK Life Sciences
8854500022
0.16 oz., 5mL Ground Glass pestle and tube 22
Each for $667.54
Only null left
K885480-0022
View Documents
DWK Life Sciences
8854800022
0.16 oz., 5mL PTFE pestle and Glass tube 22
Each for $692.18
Only null left
K885450-0025
View Documents
DWK Life Sciences
8854500025
1.96 oz., 50mL Ground Glass pestle and tube 25
Each for $910.79
Only null left
K885450-0021
View Documents
DWK Life Sciences
8854500021
0.1 oz., 3mL Ground Glass pestle and tube 21
Case for $661.89
Only null left
K885450-0023
View Documents
DWK Life Sciences
8854500023
0.59 oz., 15mL Ground Glass pestle and tube 23
Each for $761.64
Only null left
K885480-0024
View Documents
DWK Life Sciences
8854800024
1.18 oz., 30mL PTFE pestle and Glass tube 24
Each for $697.96
Only null left
K885480-0020
View Documents
DWK Life Sciences
8854800020
0.03 oz., 1mL PTFE pestle and Glass tube 20
Each for $477.07
Only null left
Description
Description
Combined conical and cylindrical surfaces improve grinding efficiency
  • Can be motor driven
  • Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
  • Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance
Specifications
Specifications
0.004 to 0.006 in.
0.1 to 0.15mm
Ideal for connective tissue such as muscle, heart and lung
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'