Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders
Combined conical and cylindrical surfaces improve grinding efficiency
Supplier:  DWK Life Sciences 8854500024
 View more versions of this product
Catalog No. K885450-0024
Only null left
Explore available promotions
Description
Description
Combined conical and cylindrical surfaces improve grinding efficiency
  • Can be motor driven
  • Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
  • Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance
Specifications
Specifications
1.18 oz.
0.1 to 0.15mm
0.39 in.
Ideal for connective tissue such as muscle, heart and lung
12.2 in.
310mm
1.25 in.
1.18 oz., 30mL
0.004 to 0.006 in.
10mm
30 mL
Ground Glass pestle and tube
8.46 in.
215mm
32mm
24
Product Suggestions
Product Suggestions