missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ 10mL SLOW-DRY™ Concentrator Tube for Evaporative Concentrators

Catalog No. K5700711025
Change view
Click to view available options
Capacity (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K5700711025 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K5700711025 Supplier DWK Life Sciences Supplier No. 5700711025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The SLOW-DRY™ concentrator tube reduces the risk of concentrating a sample to dryness.

  • Unique tip features an insulated air space around the constricted end of the tube, shielding the sample and providing a protective temperature buffer
  • Secure to assembly with 675300 polyacetal clamps
  • Tube Capacity 10mL
  • Graduation Intervals (mL) 0-1 in 0.1, 2-10 in 1
  • Height 113mm
  • Standard Taper joints 19/22
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Specifications

Product Type Concentrator Tube
For Use With (Equipment) Evaporative Concentrator
Capacity (Metric) 10 mL
Capacity (English) 0.3 oz.
Graduations 0 to 1 mL/0.1 mL, 2 to 10 mL/1 mL
Includes Slow dry tube
Standard Taper Joint 19/22
Height (English) 4.44 in.
Height (Metric) 11.3 cm