Learn More
DWK Life Sciences™ Kimble™ Kontes™ Adapter Bushing
Catalog No. K2735000022
Change view
Click to view available options
Standard Taper Joint:
10/30 Small, 24/40 Large
14/20 Small, 19/22 Large
14/35, 14/20 Small, 24/40 Large
24/40 Small, 29/42 Large
24/40 Small, 34/45 Large
24/40 Small, 45/50 Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
K2735000022 14/20 Small, 19/22 Large
K1507500266 24/40 Small, 34/45 Large
K1507500242 14/35, 14/20 Small, 24/40 Large
K1507500223 10/30 Small, 24/40 Large
K1507500268 24/40 Small, 45/50 Large
K1507500265 24/40 Small, 29/42 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. K2735000022 Supplier DWK Life Sciences™ Supplier No. 2735000022
Allow for compact assemblies of dissimiliar joints
  • Bushing type enlarging adapter features inner and outer joints for a compact way of changing joint sizes
  • Use to connect dissimilar standard taper joints
Specifications
For Use With (Application) For compact assemblies of dissimilar joints
Type Bushing Adapter
Material Glass
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Standard Taper Joint 14/20 Small, 19/22 Large