Learn More
DWK Life Sciences™ Kimble™ Kontes™ Adapter Bushing
Allow for compact assemblies of dissimiliar joints
Supplier: DWK Life Sciences™ 1507500268
Description
- Bushing type enlarging adapter features inner and outer joints for a compact way of changing joint sizes
- Use to connect dissimilar standard taper joints
Chemical Identifiers
| For compact assemblies of dissimilar joints | |
| Bushing Adapter | |
| Glass | |
| USP Type I, ASTM E438, Type I, Class A |
Specifications
| For compact assemblies of dissimilar joints | |
| Glass | |
| 24/40 Small, 45/50 Large |
| Bushing Adapter | |
| USP Type I, ASTM E438, Type I, Class A |