Learn More
DWK Life Sciences™ Kimble™ Kontes™ Adapter Bushing
Allow for compact assemblies of dissimiliar joints
Supplier:  DWK Life Sciences™ 1507500242
 View more versions of this product
Catalog No. K1507500242
Only null left
Description
Description
  • Bushing type enlarging adapter features inner and outer joints for a compact way of changing joint sizes
  • Use to connect dissimilar standard taper joints
Specifications
Chemical Identifiers
For compact assemblies of dissimilar joints
Bushing Adapter
Glass
USP Type I, ASTM E438, Type I, Class A
Specifications
For compact assemblies of dissimilar joints
Glass
14/35, 14/20 Small, 24/40 Large
Bushing Adapter
USP Type I, ASTM E438, Type I, Class A
Product Suggestions
Product Suggestions