missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kimcote™ Heavy Wall Recovery Flask
Plastic-coated single neck flask with a standard taper outer joint designed for easy recovery of reaction products
$190.80 - $190.80
Specifications
| Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
|---|---|
| Product Type | Heavy wall recovery flask |
| Socket Joint | 24/40 mm |
| Autoclavable | Autoclavable |
| Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity | |||
|
12141500
|
DWK Life Sciences
KC25165100 |
100 mL | 55 mm | 118 mm |
Case for $190.80
|
|
|||
Description
- Autoclavable KimCote™ safety coating reduces hazards of shattered glass and leakage of toxic or corrosive chemicals
- Even wall thickness and minimum runout make flasks ideal for use with rotary evaporators
- Heavy wall square joint bead
- Total contents accessible with a spatula or policeman
- Lower portion fits standard heating mantles
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
| USP Type I, ASTM E438, Type I, Class A | |
| Heavy wall recovery flask | |
| 24/40 mm | |
| Autoclavable |