missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMCOTE™ Heavy Duty Carboy
Designed for storage of solutions, and ideal for media preparation. Autoclavable safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals.
$2020.07 - $2020.07
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Closure Type | Stopper |
| Coating | Safety Coating |
| Color | Clear |
| For Use With (Application) | Solution Storage, Media Preparation |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
12-141-310
|
DWK Life Sciences
KC1495035 |
13.2 L |
Case for $2,020.07
|
|
|||||
Description
- Autoclavable safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
- With sloping shoulders of the carboy-style
- Neck is tooled for a uniform fit with a #12 rubber stopper
- 5 gallon size is designed from Federal Specification DD-B-597
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
| Autoclavable | |
| Safety Coating | |
| Solution Storage, Media Preparation | |
| Narrow | |
| Slope Shoulders, Narrow Neck |
| Stopper | |
| Clear | |
| Borosilicate Glass (3.3) | |
| Carboy | |
| 12 |