missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask

Catalog No. 01183000
Change view
Click to view available options
Capacity (Metric):
100 mL
500 mL
1000 mL
2000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01183000 100 mL
01183002 500 mL
01183003 2000 mL
12141339 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01183000 Supplier DWK Life Sciences Supplier No. KC25330100

The Kimble™ KIMAX™ KimCote™ Heavy Wall, Round Bottom Flask is a plastic-coated single neck flask useful as a rotary evaporator condensate collection flask or as a replacement for European designation KS 20/35.

  • Spherical socket joint
  • Joint Size: 35/20 mm
  • Autoclavable KimCote safety coating reduces the hazards of shattered glass and leakage of toxic or corrosive chemicals
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements

Specifications

Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Standard Taper Joint 35/20
Autoclavable Autoclavable
Sterility Non-sterile
Bottom Shape Round
Diameter (Metric) Outer 60 mm
Surface Treatment Plastic Coated