missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KIMAX™ Addition Funnel with PTFE Stopcock

Supplied with a ground glass stopper and PTFE stopcock

Supplier:  DWK Life Sciences 29040F1000

 View more versions of this product

Catalog No. 10310122


Only null left

Description

Description

  • Features gradually tapering shoulders and wide neckb which facilitate cleaning
  • Sharp separations provided by narrow bore in the body above the stopcock

Certifications

ASTM Specification E1096, Type 2 requirements

Specifications

Chemical Identifiers

Borosilicate Glass
Yes
2
USP Type I, ASTM E438 Type I
60mm
Addition Funnel

Specifications

1000 mL
USP Type I, ASTM E438 Type I
Yes
60mm
Addition Funnel
Borosilicate Glass
Tolerance: ±10mL; PTFE closure
10mL
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions