missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ 5-Place NMR Tube Washers
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
$3,153.89
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|
K8970330005
|
DWK Life Sciences
8970330005 |
5 mm dia. Tubes |
Each for $3,153.89
|
|
|||||
Description
These complete five-place washers use flexible PTFE tubing and a vacuum source (not included) to direct a power stream of wash solvent to thoroughly clean the insides of 7 and 8 in. NMR tubes.
- Two sizes available for 3 and 5mm diameter tubes
- Both sizes come with a 1L reservoir bottle
- All wetted parts are constructed of chemical-resistant PTFE or borosilicate glass
- An O-ring slips over each tube to form a vacuum-tight seal before tubes are positioned for cleaning
- With a simple twist, the wash valve can be configured to wash and then dry
- Cycles can be repeated, or cleaned tubes can be removed immediately for use
Specifications
| PTFE or Borosilicate Glass | |
| NMR Tube Washer |