missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ KG-35 Borosilicate Glass Serum Bottle, Aluminum Seal without Closure

Catalog No. p-45453
Change view
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
10310240A 10 mL
10310240E 20 mL
10310240H 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10310240A Supplier DWK Life Sciences Supplier No. 61000G10
Only null left

Autoclavable Kimble™ serum bottles and vials are well-suited for the handling, containment and storage of a variety of liquids including reagents, vaccines, blood plasma, culture media, chromatography samples and more.

  • Ideal for packaging and storage where applications of injectable and parenteral solutions require utmost purity
  • Highly resistant to thermal and mechanical shock as well as chemical attack
  • Manufactured from USP Type I borosilicate molded glass

Specifications

Capacity 0.33 oz., 10mL
Capacity (Metric) 10 mL
Capacity (English) 0.33 oz.
Diameter (English) 0.98 in.
Material Borosilicate Glass
Length (Metric) 52mm
Product Type Serum Bottle
Length (English) 2.04 in.
Closure Material Aluminum
Closure Size 20A
Diameter (Metric) 25 mm
Show More Show Less