Learn More
DWK Life Sciences Kimble™ Disposable Micro Capillary Pipets
Color-coded for selection of correct size
 View more versions of this product
Catalog No. 13-678-18B
Only null left
Explore available promotions
Description
Description
Color-coded for selection of correct size
  • Length: 5 in. with single black capacity ring
  • Clean and ready to use
  • To dispense, simply cover hole on rubber bulb and squeeze
Order Info
  • Packaged in easy open cylinders for convenient dispensing.
  • Each cylinder contains 250 pipets and one pipet device.
Compliance
Calibrated To Contain. Made of soda lime glass in compliance with ASTM™ Specification E438, Type II glass requirements.
Specifications
Specifications
ASTM , E672, USP type III
5 in.
Black
127 mm
Soda Lime Glass
±0.5 μL
20 μL
Microcapillary Pipet
Product Suggestions
Product Suggestions