missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Disposable Micro Capillary Pipets

Numéro de catalogue. p-163209
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
50 μL
100 μL
Color:
Black
Blue
Green
Orange
White
Tolerance:
±0.5 μL
±1 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric) Color Tolerance
K7190050 50 μL Green ±0.5 μL
1367818B 20 μL Black ±0.5 μL
1367818A 10 μL Orange ±0.5 μL
K71900100 100 μL Blue ±0.5 μL
K719005 5 μL White ±1 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. K7190050
Il en reste null

Color-coded for selection of correct size

  • Length: 5 in. with single black capacity ring
  • Clean and ready to use
  • To dispense, simply cover hole on rubber bulb and squeeze

Order Info

  • Packaged in easy open cylinders for convenient dispensing.
  • Each cylinder contains 250 pipets and one pipet device.

Compliance

Calibrated To Contain. Made of soda lime glass in compliance with ASTM™ Specification E438, Type II glass requirements.

Spécifications

Certifications/Compliance ASTM , E672, USP type III
Material Soda Lime Glass
Length (English) 5 in.
Tolerance ±0.5 μL
Color Green
Volume (Metric) 50 μL
Length (Metric) 127 mm
Type Microcapillary Pipet
Disposable Single-use