Learn More
DWK LIFE SCIENCES™ KIMBLE™ Closures for Sample Vials
Description
Black phenolic closure with PTFE-lined cap
- Compatible with media bottles and screw-thread vials
- Black phenolic material (wood filled) is highly resistant to the effects of temperature and steam encountered in autoclaving
- Cemented-in PTFE-faced rubber liner provides a reliable seal
- Available in multiple GPI thread sizes to suit a range of vial formats
Specifications
Specifications
| Closure Size | 15-425 |
| Liner Material | PTFE, Rubber |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Phenolic |
| Color | Black |
| Sterility | Non-sterile |