missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Clear Glass French Squares with Pulp/Vinyl-lined Caps

Wide mouth design for efficient addition or removal of contents

Supplier:  DWK Life Sciences 5610843V21

 View more versions of this product

Catalog No. 13756304


Only null left

Description

Description

  • French square bottles are Ideal for mixing, storing and sampling liquids or solid matter
  • Square shape maximizes use of shelf space
  • Clear glass allows for easy viewing of contents

Specifications

Chemical Identifiers

Clear
Black
Phenolic
Soda Lime Glass
Pulp/Vinyl
Screw Cap

Specifications

8 oz., 250 mL
8 oz.
2.2 in.
Convenience Pk. (cap attached)
137 mm
5.39 in.
Phenolic
56 mm
250 mL
Clear
Soda Lime Glass
Black
French Square Bottle
Pulp/Vinyl
43-400 mm
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions