missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences KIMBLE™ KONTES™ Support Frit for HPLC 90mm Ultra-Ware™ Conversion Base
Description
- Coarse porosity glass frit designed for HPLC applications
- Compatible with 90mm Ultra-Ware Conversion Base
- Replacement support frit for maintaining filtration performance
- Supplied by DWK Life Sciences under the Kimble Kontes brand
Specifications
Specifications
| Disposable | Reusable |
| Material | Glass |
| Sterility | Non-sterile |
| Diameter (Metric) Inside | 90 mm |
| For Use With (Equipment) | Filtration Assembly and vacuum pump |