missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Storage Cases for MICROFLEX™ Kits

Catalog No. K7460010001
Change view
Click to view available options
Product Type:
Storage Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
K7460010001 Storage Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K7460010001 Supplier DWK Life Sciences Supplier No. 7460010001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Component for standard microscale kits

  • Measures: 2.5 x 13.5 x 10 in. (6.4 x 34.3 x 25.4cm)

Specifications

For Use With (Equipment) For standard microscale kit, threaded standard 14/10 kit
Product Type Storage Case