Learn More
DWK Life Sciences Kimble™ Kontes™ Rotary Evaporator Traps With Inner Vapor Tube at Top
Design allows for expanded volume for mixtures that have a tendency to foam excessively
$603.46 - $686.07
Specifications
Certifications/Compliance USP Type I and ASTM E438, Class A
Material Borosilicate Glass
Product Type Rotary Evaporator Trap
For Use With (Application) Provides for expanded volume for organic mixtures that have a tendency to foam excessively
Includes Inner vapor tube at top
View More Specs
Products 7
Catalog Number Mfr. No. Standard Taper Joint Capacity (English) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Capacity (English) Height (English) Price Quantity & Availability  
K5702100114
View Documents Promotion Details
DWK Life Sciences
5702100114
Upper: 24/40, Lower: 14/20 3.4 oz. 6.6 in.
Each for $603.46
Only null left
K5702100119
View Documents Promotion Details
DWK Life Sciences
5702100119
Upper: 24/40, Lower: 19/22 3.4 oz. 6.6 in.
Each for $603.46
Only null left
K5702100124
View Documents Promotion Details
DWK Life Sciences
5702100124
Upper: 24/40, Lower: 24/40 3.4 oz. 7.4 in.
Each for $649.71
Only null left
K5702102514
View Documents Promotion Details
DWK Life Sciences
5702102514
Upper: 24/40, Lower: 14/20 8.45 oz. 7.4 in.
Each for $621.04
Only null left
K5702102519
View Documents Promotion Details
DWK Life Sciences
5702102519
Upper: 24/40, Lower: 19/22 8.45 oz. 7.4 in.
Each for $655.57
Only null left
K5702102524
View Documents Promotion Details
DWK Life Sciences
5702102524
Upper: 24/40, Lower: 24/40 8.45 oz. 8.2 in.
Each for $678.82
Only null left
K5702105024
View Documents Promotion Details
DWK Life Sciences
5702105024
Upper: 24/40, Lower: 24/40 16.9 oz. 9 in.
Each for $686.07
Only null left
Description
Description
  • Inner vapor tube is ring-sealed at top of flask to prevent entry of organic mixtures into the condenser
  • Flasks have standard taper 24/40 outer ground joint at top
Specifications
Specifications
USP Type I and ASTM E438, Class A
Rotary Evaporator Trap
Inner vapor tube at top
Borosilicate Glass
Provides for expanded volume for organic mixtures that have a tendency to foam excessively