Learn More
DWK Life Sciences Kimble™ Kontes™ Rotary Evaporator Traps With Inner Vapor Tube at Bottom
Use when foam is likely to be created and drawn into the condenser
Supplier:  DWK Life Sciences 5702000114
 View more versions of this product
Catalog No. K5702000114
Only null left
Description
Description
  • Flask has a standard taper 24/40 outer ground joint at the top
Specifications
Chemical Identifiers
USP Type I and ASTM E438, Class A
Rotary Evaporator Trap
Inner vapor tube at bottom
Borosilicate Glass
Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping
Specifications
Upper: 24/40, Lower: 14/20
6.25 in.
100mL
3.4 oz., 100mL
USP Type I and ASTM E438, Class A
Inner vapor tube at bottom
3.4 oz.
16cm
Borosilicate Glass
Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping
Rotary Evaporator Trap
Product Suggestions
Product Suggestions