Learn More
DWK Life Sciences Kimble™ Kontes™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
For use with screw-thread culture and centrifuge tubes
Supplier:  DWK Life Sciences 7380815415
 View more versions of this product
Catalog No. 0334077BB
Only null left
Description
Description
  • Provides superior closure system capable of maintaining its liner through multiple autoclavable operations
  • PTFE-coated surface of exposed liner is chemical-resistant and compatible with sensitive, corrosive and volatile solvents
Specifications
Chemical Identifiers
Screw Cap
Autoclavable
EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes
PTFE, Rubber
Polypropylene
Specifications
15-415
Screw Cap
Polypropylene
EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes
PTFE, Rubber
Autoclavable
For repeated autoclaving, to provide superior closure system
Product Suggestions
Product Suggestions