missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Pear-Shaped Heavy-Wall Boiling Flasks

Catalog No. K6087002524
Change view
Click to view available options
Boiling Flask, Capacity: 250mL, Joint: 24/40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
K6087002524 Boiling Flask, Capacity: 250mL, Joint: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K6087002524 Supplier DWK Life Sciences Supplier No. 6087002524

Single neck flask with a standard outer joint at top

  • Capacity: 250mL
  • Joint: 24/40
  • Single neck

Specifications

Capacity (Metric) 250 mL
Material Borosilicate Glass
Standard Taper Joint 24/40