missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Kimble™ Kontes™ MIDI-VAP 4000 Component Replacement Parts

Component parts for both ammonia and phenol distillation system

Supplier:  DWK Life Sciences™ 4794550050

 View more versions of this product

Catalog No. K4794550050


Only null left

Description

Description

Component parts for both ammonia and phenol distillation system

  • For use with MIDI-VAP 4000 Distillation Apparatus
Specifications

Chemical Identifiers

Distillation systems

Specifications

All fittings
Reaction/Absorption Tube
50mL
29/42
Distillation systems
1.6 oz., 50mL
MIDI-VAP 4000 Cyanide System
1.6 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions