missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Kimble™ Kontes™ MIDI-VAP 4000 Component Replacement Parts

Catalog No. K4104800014
Change view
Click to view available options
Type:
Ammonia Phenol Glassware 1 Set
Ammonia Phenol Glassware 5 Sets
Cyanide Distillation
Cyanide Glassware Set
Distilling Head
Long Stem Outlet Tube
Manifold Heater
Open Top Screw Thread Cap
Quick Disconnect Fittings, Female
Quick Disconnect Fittings, Male
Reaction Tube
Reaction/Absorption Tube
Receiver Tube
Sealing Ring
Short Stem Outlet Tube
Tubing Kit
For Use With (Equipment):
For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
MIDI-VAP 4000
MIDI-VAP 4000 Ammonia System
MIDI-VAP 4000 Ammonia/Phenol System
MIDI-VAP 4000 Cyanide System
MIDI-VAP 4000 Phenol System
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
K4104800014 Sealing Ring For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
K4104790014 Open Top Screw Thread Cap MIDI-VAP 4000 Ammonia/Phenol System
K4794550050 Reaction/Absorption Tube MIDI-VAP 4000 Cyanide System
K4104810014 Cyanide Distillation For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
K4794564501 Distilling Head MIDI-VAP 4000 Ammonia/Phenol System
K4794580001 Long Stem Outlet Tube MIDI-VAP 4000 Ammonia System
K4794590005 Ammonia Phenol Glassware 5 Sets MIDI-VAP 4000 Ammonia/Phenol System
K4794590000 Ammonia Phenol Glassware 1 Set MIDI-VAP 4000 Ammonia/Phenol System
K4794580000 Short Stem Outlet Tube MIDI-VAP 4000 Phenol System
K4794600005 Cyanide Glassware Set MIDI-VAP 4000
K4794610000 Distilling Head MIDI-VAP 4000 Cyanide System
K4794700050 Reaction Tube MIDI-VAP 4000 Ammonia/Phenol System
K7204404000 Manifold Heater MIDI-VAP 4000
K4794710050 Receiver Tube MIDI-VAP 4000 Ammonia/Phenol System
K4794000014 Quick Disconnect Fittings, Female For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
K4794000022 Cyanide Distillation MIDI-VAP 4000
K4794000018 Quick Disconnect Fittings, Male For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
K4794180046 Tubing Kit MIDI-VAP 4000
K4794180043 Tubing Kit MIDI-VAP 4000 Ammonia/Phenol System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. K4104800014 Supplier DWK Life Sciences™ Supplier No. 4104800014

Component parts for both ammonia and phenol distillation system

Component parts for both ammonia and phenol distillation system

  • For use with MIDI-VAP 4000 Distillation Apparatus

Specifications

For Use With (Application) Distillation systems
Type Sealing Ring
Material PTFE/Silicone Liner
For Use With (Equipment) For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System