Learn More
DWK Life Sciences Kimble™ Kontes™ Microscale B - Connector
Unique design adapts GPI threaded glass to a serrated PTFE hose connection or 0.25 in. -28 tubing connections
Supplier: DWK Life Sciences 7471850013
Description
- Fits hose I.D.: 0.25 in.
- Internal Thread Size: 0.25 in.-28
- GPI Thread Size: 13-425
Specifications
| PTFE | |
| 13-425 | |
| Hose Connector |