Learn More
DWK Life Sciences Kimble™ Kontes™ Microflex™ Microscale Kits - Inlet/Vacuum Adapter
Compression cap has Viton™ O-ring
Supplier:  DWK Life Sciences 7460350814
Catalog No. K746035-0814
Only null left
Explore available promotions
Description
Description
  • Use in Cold Finger Condenser (22-300-624) or Sublimation Assembly Condenser (K746125-0000), or other plain-stemmed glassware with O.D. from 3/16 to 5/16 in. (5 to 8mm)
Specifications
Specifications
For Microflex™ microscale kits, cold finger condenser or sublimation assembly condense or other plain-stemmed glassware with O.D. from 0.18 to 0.31 in., fits tubing I.D. 0.25 in.
Glass
14/10
Hose connection
15-415
thermometer Inlet Adapter
Product Suggestions
Product Suggestions