Learn More
DWK Life Sciences KIMBLE™ KONTES™ HPLC 90mm Ultra-Ware Conversion Base
Catalog No. K9539840090
Change view
Click to view available options
Material:
Glass, PTFE
Catalog No. Material
K9539840090 Glass, PTFE
1 options
Catalog No. K9539840090 Supplier DWK Life Sciences Supplier No. 9539840090
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The KIMBLE™ KONTES™ HPLC 90mm Ultra-Ware Conversion Base converts Five Valve, Four Valve, and Filtration caps to use 90mm filter membranes, enabling up to four times faster filtration. Made from glass-filled PTFE; supplied with a coarse glass frit and screw clamp.
  • Converts Five Valve, Four Valve, and Filtration ULTRA-WARE caps to accept 90 mm filter membranes
  • Provides up to four times faster filtration compared to standard membrane sizes
  • Manufactured from glass-filled PTFE for chemical resistance
  • Supplied with a coarse porosity glass support frit
  • Includes an upper screw clamp for secure assembly
  • Compatible with Standard ULTRA-WARE Recirculation/Filtration cap systems
Specifications
Disposable Reusable
Material Glass, PTFE
Sterility Non-sterile
For Use With (Equipment) Bottle or reservoir with a GL 45 thread