Learn More
DWK Life Sciences Kimble™ Kontes™ Extraction Manifold Replacement Components
For extraction manifold.
$354.86 - $431.11
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|
K9538110000
|
DWK Life Sciences
9538110000 |
PTFE Gasket |
Pack of 25 Each for $354.86
|
|
|||||
|
K9711010347
|
DWK Life Sciences
9711010347 |
Glass Funnel (300 mL) |
Each for $431.11
|
|
|||||
Description
- For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
- Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
Specifications
| Solid Phase Extraction Manifold |