Learn More
DWK Life Sciences™ Kimble™ Kontes™ Distillation Apparatus Components: Hex Head Stopper
Catalog No. K8508002440
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
K8508002440 24/40
1 options
Catalog No. K8508002440 Supplier DWK Life Sciences™ Supplier No. 8508002440
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
  • Hollow, hexagonal head, Standard Taper stopper
  • May be set on the flattened top to avoid contamination of the ground surface
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Type Hex Head Stopper
Material Glass
For Use With (Equipment) Distillation apparatus
Standard Taper Joint 24/40