Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Accessories
Catalog No. K7473000100
Change view
Click to view available options
Product Type:
Air Condenser
Drying Tube/Chromatography Column
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
K7473000100 Air Condenser
K7473000050 Drying Tube/Chromatography Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K7473000100 Supplier DWK Life Sciences Supplier No. 7473000100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For chromatography columns
  • 747000 Series Microflex Kits
  • Features versatile threaded chaste connections
  • Can be assembled rightside up or upside down, providing great versatility
  • GPI thread size: 13 to 425
Specifications
For Use With (Equipment) For chromatography columns
Thread Size 13-425
Product Type Air Condenser