Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Accessories
For chromatography columns
Supplier:  DWK Life Sciences 7473000050
 View more versions of this product
Catalog No. K7473000050
Only null left
Description
Description
  • 747000 Series Microflex Kits
  • Features versatile threaded chaste connections
  • Can be assembled rightside up or upside down, providing great versatility
  • GPI thread size: 13 to 425
Specifications
Chemical Identifiers
For chromatography columns
13-425
Specifications
For chromatography columns
13-425
Drying Tube/Chromatography Column
Product Suggestions
Product Suggestions