Learn More
DWK Life Sciences KIMBLE™ KONTES™ 90mm Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support
Catalog No. K9537520090
Change view
Click to view available options
Material:
Fritted Glass
Catalog No. Material
K9537520090 Fritted Glass
1 options
Catalog No. K9537520090 Supplier DWK Life Sciences Supplier No. 9537520090
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KIMBLE™ KONTES™ 90mm Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support is designed for laboratory microfiltration applications, featuring a 90mm filter size and fritted glass base support for reliable sample processing.
  • 90mm filter size suitable for standard microfiltration workflows
  • Fritted glass support base provides stable, uniform filtration
  • Ultra-Ware™ construction for chemical resistance and durability
  • Compatible with standard 90mm membrane filters
  • Designed for use in laboratory vacuum filtration assemblies
Specifications
Disposable Reusable
Material Fritted Glass
Sterility Non-sterile
For Use With (Equipment) Filtration Assembly and vacuum pump