Learn More
DWK Life Sciences™ Kimble™ Sample Vials with Black Phenolic Caps and Polyvinyl-faced Pulp Liners
Kimble™ Screw thread N-51A Borosilicate Glass Sample Vials made of low extractable Kimax N-51A glass.
$120.65 - $1,819.11
Specifications
Closure Color Black
Color Clear
Closure Material Phenolic
View More Specs
Products 10
Catalog Number Mfr. No. Volume (Metric) Closure Size Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Closure Size Diameter (Metric) Outer Price Quantity & Availability  
0334015
View Documents Promotion Details
DWK Life Sciences™
60910C12
2 mL 8-425 12 mm
Pack of 144 Each for $132.51
Case of 16 Pack for $2,120.16
Only null left
0334016
View Documents Promotion Details
DWK Life Sciences™
60910C1
4 mL 13-425 15 mm
Pack of 144 Each for $224.07
Case of 16 Pack for $3,585.12
Only null left
0334017
View Documents Promotion Details
DWK Life Sciences™
60910C112
5.5 mL 13-425 16 mm
Pack of 144 Each for $182.70
Case of 16 Pack for $2,923.20
Only null left
0334018
View Documents Promotion Details
DWK Life Sciences™
60910C2
8 mL 15-425 17 mm
Pack of 144 Each for $120.65
Case of 12 Pack for $1,447.80
Only null left
0333938F
View Documents Promotion Details
DWK Life Sciences™
60910C4
16 mL 18-400 21 mm
Case of 1152 Each for $1,819.11
Only null left
11417230
View Documents Promotion Details
DWK Life Sciences™
60957C4
16 mL 24-400 28 mm
Case of 432 Each for $990.96
Only null left
0334021
View Documents Promotion Details
DWK Life Sciences™
60910C6
24 mL 20-400 23 mm
Pack of 144 Each for $242.14
Case of 6 Pack for $1,452.84
Only null left
17000617
View Documents Promotion Details
DWK Life Sciences™
DWK60957C6
24 mL 24-400 24 mm
Case of 432 Each for $913.38
Only null left
0334022
View Documents Promotion Details
DWK Life Sciences™
60910C8
30 mL 22-400 25 mm
Pack of 144 Each for $285.70
Case of 4 Pack for $1,142.80
Only null left
11417130
View Documents Promotion Details
DWK Life Sciences™
60957C11
41 mL 24-400 28 mm
Case of 432 Each for $1,304.43
Only null left
Description
Description
Kimble™ Screw-Thread N-51A Borosilicate Glass Sample Vials made of low extractable Kimax N-51A glass.
  • Vials with phenolic caps and polyvinyl-faced liners
  • Borosilicate glass sample vials with caps
  • Caps are not attached
  • Clear screw thread sample vial
  • Closed-top black phenolic closure with polyvinyl-faced pulp liner (not attached)
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Compliance
Kimax N-51A glass conforms to ASTM Type 1, Class B requirements.
Specifications
Specifications
Black
Clear
Phenolic