missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Storage/Media Bottles

With linerless blue polypropylene caps
$639.18 - $1,954.68
Specifications
| Color | Ultra Clear |
|---|---|
| Diameter (English) Inner Neck | 1.18 in. |
| Diameter (Metric) Inner Neck | 30 mm |
| Includes | Clear drip-free Polypropylene pouring ring |
| Material | Plastic |
| Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
12141320
![]() |
DWK Life Sciences
KC14395100 |
100 mL | 20 to 80mL |
Case of 4 Each for $639.18
|
|
||||
12141321
![]() |
DWK Life Sciences
KC14395250 |
250 mL | 50 to 200mL |
Case of 4 Each for $724.18
|
|
||||
12141322
![]() |
DWK Life Sciences
KC14395500 |
500 mL | 100 to 400mL |
Case of 4 Each for $935.82
|
|
||||
12141323
![]() |
DWK Life Sciences
KC143951000 |
1000 mL | 100 to 900mL |
Case of 4 Each for $1,148.59
|
|
||||
12141324
![]() |
DWK Life Sciences
KC143952000 |
2000 mL | 400 to 1800mL |
Case of 4 Each for $1,954.68
|
|
||||
|
12141325
|
DWK Life Sciences
KC143955000 |
5000 mL | 1000 to 4000mL |
Case for $1,124.18
|
|
||||
|
12141326
|
DWK Life Sciences
KC1439510000 |
10,000 mL | 2000 to 8000mL |
Case for $1,726.18
|
|
||||
Description
With linerless blue polypropylene caps
- KimCote™ plastic coating helps prevent and contain spills in the event of glassware breakage
- GL 45 screw thread bottles with white graduations and marking spots
- Wide opening for easy access (approx. 30mm I.D.)
- Linerless cap is blue polypropylene and autoclavable (140°C max)
- Replaceable pour ring included
- Ultra-clear for improved sample viewing
- Non-slip surface — wet or dry
Applications:
Pressurized glass, chemical storage, transportation of samples, handling of hazardous chemicals, harmful biologicals and acids
Specifications
| Ultra Clear | |
| 30 mm | |
| Plastic | |
| Yes | |
| GL 45 |
| 1.18 in. | |
| Clear drip-free Polypropylene pouring ring | |
| Blue | |
| Polypropylene | |
| Screw Cap |